3-Bromopyridine-2-thiol 98%
Leverantör: Apollo Scientific
Synonyms:
3-Bromopyridine-2(1H)-thione
OR901714-1G
OR901714-5G
APOSOR901714-1GEA
2575
SEK
APOSOR901714-1G
APOSOR901714-5G
3-Bromopyridine-2-thiol 98%
3-Bromopyridine-2-thiol
Formel:
C₅H₄BrNS MW: 190,061 g/mol Storage Temperature: Omgivande |
MDL Number:
MFCD14651478 CAS nummer: 65938-86-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...