4-(1,3,2-Dioxaborinan-2-yl)phenol 98%
Leverantör: Apollo Scientific
OR901581-1G OR901581-5G
APOSOR901581-1GEA 1025 SEK
APOSOR901581-1G APOSOR901581-5G
4-(1,3,2-Dioxaborinan-2-yl)phenol 98%
4-(1,3,2-Dioxaborinan-2-yl)phenol
Formel: C₉H₁₁BO₃
MW: 177,993 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD21609692
CAS nummer: 1640035-73-9