(5-Methoxycarbonyl-1-methylpyrrol-2-yl)boronic acid, pinacol ester 96%
Leverantör: Apollo Scientific
OR901449-1G OR901449-5G
APOSOR901449-1GEA 3550 SEK
APOSOR901449-1G APOSOR901449-5G
(5-Methoxycarbonyl-1-methylpyrrol-2-yl)boronic acid, pinacol ester 96%
(5-Methoxycarbonyl-1-methylpyrrol-2-yl)boronic acid, pinacol ester
Formel: C₁₃H₂₀BNO₄
MW: 265.113 g/mol
Storage Temperature: Frys
MDL Number: MFCD29035415
CAS nummer: 2377607-43-5