3,4-Diethylpyrrole 98%

Leverantör: Apollo Scientific

OR900851-1G OR900851-5G
APOSOR900851-1GEA 5650 SEK
APOSOR900851-1G APOSOR900851-5G
3,4-Diethylpyrrole 98%
3,4-Diethylpyrrole
Formel: C₈H₁₃N
Storage Temperature: Omgivande
MDL Number: MFCD08062571
CAS nummer: 16200-52-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR