Methyl 5-formylpyrrole-2-carboxylate 98%
Leverantör: Apollo Scientific
Synonyms: Methyl 5-formyl-1H-pyrrole-2-carboxylate
OR900846-10G OR900846-1G OR900846-5G
APOSOR900846-10GEA 6775 SEK
APOSOR900846-10G APOSOR900846-1G APOSOR900846-5G
Methyl 5-formylpyrrole-2-carboxylate 98%
Methyl 5-formylpyrrole-2-carboxylate
Formel: C₇H₇NO₃
MW: 153,135 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD09878545
CAS nummer: 1197-13-3