4-(S-Acetylthio)benzaldehyde 95%
Leverantör: Apollo Scientific
OR900844-1G
OR900844-500MG
APOSOR900844-1GEA
9500
SEK
APOSOR900844-1G
APOSOR900844-500MG
4-(S-Acetylthio)benzaldehyde 95%
4-(S-Acetylthio)benzaldehyde
Formel:
C₉H₈O₂S MW: 180,224 g/mol Storage Temperature: Omgivande |
MDL Number:
MFCD09260458 CAS nummer: 28130-89-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...