(3S,4S)-Tetrahydrofuran-3,4-diol
Leverantör: Apollo Scientific
|
Varning
|
Synonyms:
L-Threitan, 1,4-Anhydro-L-threitol
OR8886-1G
OR8886-250MG
APOSOR8886-1GEA
14900
SEK
APOSOR8886-1G
APOSOR8886-250MG
(3S,4S)-Tetrahydrofuran-3,4-diol
(3S,4S)-Tetrahydrofuran-3,4-diol
Formel:
C₄H₈O₃ MW: 104,105 g/mol Storage Temperature: Kylskåp |
MDL Number:
MFCD00075211 CAS nummer: 84709-85-3 UN: 1325 ADR: 4.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...