1H-Indazole-5-boronic acid, pinacol ester, N1-BOC protected
Leverantör: Apollo Scientific
Varning
Synonyms: 1-(Tert-butoxycarbonyl)-5-(4,4,5,5-Tetramethyl-[1,3,2]Dioxaborolan-2-yl)indazole, 1-Boc-1H-indalzole-5-boronic acid pinacol ester, tert-Butyl 5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indazole-1-carboxylate, 1-(tert-Butoxycarbonyl)-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indazole
OR8862-1G OR8862-250MG
APOSOR8862-1GEA 1200 SEK
APOSOR8862-1G APOSOR8862-250MG
1H-Indazole-5-boronic acid, pinacol ester, N1-BOC protected
1H-Indazole-5-boronic acid, pinacol ester, N1-BOC protected
Formel: C₁₈H₂₅BN₂O₄
MW: 344,213 g/mol
Smältpunkt: 110…113 °C
Storage Temperature: Omgivande
MDL Number: MFCD08437629
CAS nummer: 864771-44-8
UN: 2811
ADR: 6.1,III