6-(Benzyloxy)-5-methoxy-1H-indole-2-carboxylic acid
Leverantör: Apollo Scientific
Varning
Synonyms: NSC92530, 6-(Benzyloxy)-2-carboxy-5-methoxy-1H-indole
OR8600T-1G OR8600T-250MG
APOSOR8600T-1GEA 11050 SEK
APOSOR8600T-1G APOSOR8600T-250MG
6-(Benzyloxy)-5-methoxy-1H-indole-2-carboxylic acid
6-(Benzyloxy)-5-methoxy-1H-indole-2-carboxylic acid
Formel: C₁₇H₁₅NO₄
MW: 297,305 g/mol
Smältpunkt: 230…232 °C (dec.)
Storage Temperature: Frys
MDL Number: MFCD00055977
CAS nummer: 2495-92-3