5,6-Dimethyl-2-(pyridin-2-yl)pyrimidin-4-ol
Leverantör: Apollo Scientific
Varning
Synonyms: 2-(4,5-Dimethyl-6-hydroxypyrimidin-2-yl)pyridine, 4,5-Dimethyl-6-hydroxy-2-(pyridin-2-yl)pyrimidine
OR8472-1G OR8472-5G
APOSOR8472-1GEA 3450 SEK
APOSOR8472-1G APOSOR8472-5G
5,6-Dimethyl-2-(pyridin-2-yl)pyrimidin-4-ol
5,6-Dimethyl-2-(pyridin-2-yl)pyrimidin-4-ol
Formel: C₁₁H₁₁N₃O
MW: 201,225 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00269287
CAS nummer: 204394-52-5