4-[4-(2-Methoxyphenyl)piperidino]-3-nitrobenzaldehyde 97%

Leverantör: Apollo Scientific

OR8110-1G OR8110-5G
APOSOR8110-1GEA 1325 SEK
APOSOR8110-1G APOSOR8110-5G
4-[4-(2-Methoxyphenyl)piperidino]-3-nitrobenzaldehyde 97%
4-[4-(2-Methoxyphenyl)piperidino]-3-nitrobenzaldehyde
Formel: C₁₉H₂₀N₂O₄
Smältpunkt: 116…118 °C
Storage Temperature: Omgivande
MDL Number: MFCD01314135
CAS nummer: 301334-92-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR