4-Chlorophenylcyanamide

Leverantör: Apollo Scientific

OR7950T-250MG OR7950T-25MG
APOSOR7950T-250MGEA 9875 SEK
APOSOR7950T-250MG APOSOR7950T-25MG
4-Chlorophenylcyanamide
4-Chlorophenylcyanamide
Formel: C₇H₅ClN₂
MW: 152.581 g/mol
Smältpunkt: 103…105 °C
Storage Temperature: Omgivande
MDL Number: MFCD00114383
CAS nummer: 13463-94-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR