5-Bromo-N-(2,2-dimethoxyethyl)pyrimidin-2-amine
Leverantör: Apollo Scientific
OR7850-1G OR7850-5G
APOSOR7850-1GEA 520 SEK
APOSOR7850-1G APOSOR7850-5G
5-Bromo-N-(2,2-dimethoxyethyl)pyrimidin-2-amine
5-Bromo-N-(2,2-dimethoxyethyl)pyrimidin-2-amine
Formel: C₈H₁₂BrN₃O₂
Storage Temperature: Omgivande
MDL Number: MFCD03646021
CAS nummer: 885267-37-8