3'-Methyl-[1,1'-biphenyl]-2-carboxylic acid
Leverantör: Apollo Scientific
Varning
Synonyms: 2-Carboxy-3'-methylbiphenyl, 2-(3-Methylphenyl)benzoic acid
OR7516-1G OR7516-5G
APOSOR7516-1GEA 1475 SEK
APOSOR7516-1G APOSOR7516-5G
3'-Methyl-[1,1'-biphenyl]-2-carboxylic acid
3'-Methyl-[1,1'-biphenyl]-2-carboxylic acid
Formel: C₁₄H₁₂O₂
MW: 212,244 g/mol
Smältpunkt: 94…97 °C
Storage Temperature: Omgivande
MDL Number: MFCD04039113
CAS nummer: 107412-71-5