3'-Chloro-[1,1'-biphenyl]-4-carboxaldehyde 95%
Leverantör: Apollo Scientific
Varning
OR7349-1G OR7349-5G
APOSOR7349-1GEA 940 SEK
APOSOR7349-1G APOSOR7349-5G
3'-Chloro-[1,1'-biphenyl]-4-carboxaldehyde 95%
3'-Chloro-[1,1'-biphenyl]-4-carboxaldehyde
Formel: C₁₃H₉ClO
Storage Temperature: Omgivande
MDL Number: MFCD03424617
CAS nummer: 400744-49-2