3'-Chloro-[1,1'-biphenyl]-2-carboxaldehyde
Leverantör: Apollo Scientific
Varning
OR7347-1G OR7347-500MG
APOSOR7347-1GEA 7375 SEK
APOSOR7347-1G APOSOR7347-500MG
3'-Chloro-[1,1'-biphenyl]-2-carboxaldehyde
3'-Chloro-[1,1'-biphenyl]-2-carboxaldehyde
Formel: C₁₃H₉ClO
MW: 216,663 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD03424628
CAS nummer: 216443-25-3