4-Formylbenzeneboronic acid, propane-1,3-diol cyclic ester
Leverantör: Apollo Scientific
Varning
OR7217-1G OR7217-500MG
APOSOR7217-1GEA 585 SEK
APOSOR7217-1G APOSOR7217-500MG
4-Formylbenzeneboronic acid, propane-1,3-diol cyclic ester
4-Formylbenzeneboronic acid, propane-1,3-diol cyclic ester
Formel: C₁₀H₁₁BO₃
MW: 190,004 g/mol
Smältpunkt: 72…73 °C
Storage Temperature: Omgivande
MDL Number: MFCD04039026
CAS nummer: 4463-41-6