3,4-Dihydro-2H-1,4-oxazine, N-BOC protected

Leverantör: Apollo Scientific
Varning

Synonyms: tert-Butyl 2,3-dihydro-4H-1,4-oxazine-4-carboxylate, 4-(tert-Butoxycarbonyl)-3,4-dihydro-2H-1,4-oxazine

OR70142-1G OR70142-5G
APOSOR70142-1GEA 1475 SEK
APOSOR70142-1G APOSOR70142-5G
3,4-Dihydro-2H-1,4-oxazine, N-BOC protected
3,4-Dihydro-2H-1,4-oxazine, N-BOC protected
Formel: C₉H₁₅NO₃
MW: 185,22 g/mol
Kokpunkt: 46…47 °C (0,225 mm Hg)
Storage Temperature: Omgivande
MDL Number: MFCD16039461
CAS nummer: 1221347-27-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR