1-[3-(Piperidin-1-yl)prop-1-yl]piperazine 98%
Leverantör: Apollo Scientific
Varning
Synonyms: 1-[3-(Piperazin-1-yl)prop-1-yl]piperidine
OR6892-10G OR6892-2G OR6892-5G
APOSOR6892-10GEA 6000 SEK
APOSOR6892-10G APOSOR6892-2G APOSOR6892-5G
1-[3-(Piperidin-1-yl)prop-1-yl]piperazine 98%
1-[3-(Piperidin-1-yl)prop-1-yl]piperazine
Formel: C₁₂H₂₅N₃
MW: 211,347 g/mol
Kokpunkt: 116…118 °C
Storage Temperature: Omgivande
MDL Number: MFCD01320896
CAS nummer: 111594-93-5