trans-4-Cyanocinnamic acid 98%
Leverantör: Apollo Scientific
Varning
Synonyms: (2E)-3-(4-Cyanophenyl)prop-2-enoic acid, (E)-3-(4-Cyanophenyl)acrylic acid
OR6585-1G OR6585-500MG
APOSOR6585-1GEA 310 SEK
APOSOR6585-1G APOSOR6585-500MG
trans-4-Cyanocinnamic acid 98%
trans-4-Cyanocinnamic acid
Formel: C₁₀H₇NO₂
MW: 173,168 g/mol
Smältpunkt: 245…248 °C
Storage Temperature: Omgivande
MDL Number: MFCD00016845
CAS nummer: 16642-94-7