trans-3,4-Dimethoxycinnamic acid 98%
Leverantör: Apollo Scientific
Varning
Synonyms: (E)-3-(3,4-Dimethoxyphenyl)acrylic acid, (2E)-3-(3,4-Dimethoxyphenyl)prop-2-enoic acid, (E)-3,4-Dimethoxycinnamic acid
OR6575-25G OR6575-5G
APOSOR6575-25GEA 395 SEK
APOSOR6575-25G APOSOR6575-5G
trans-3,4-Dimethoxycinnamic acid 98%
trans-3,4-Dimethoxycinnamic acid
Formel: C₁₁H₁₂O₄
MW: 208,211 g/mol
Smältpunkt: 177…179 °C
Storage Temperature: Omgivande
MDL Number: MFCD00004387
CAS nummer: 14737-89-4
EINECS: 219-025-5