5-Chloro-3-cyanopyridine-2-carboxylic acid

Leverantör: Apollo Scientific
Varning

Synonyms: 5-Chloro-3-cyanopicolinic acid, 2-Carboxy-5-chloronicotinonitrile

OR61368-1G OR61368-500MG
APOSOR61368-1GEA 14350 SEK
APOSOR61368-1G APOSOR61368-500MG
5-Chloro-3-cyanopyridine-2-carboxylic acid
5-Chloro-3-cyanopyridine-2-carboxylic acid
Formel: C₇H₃ClN₂O₂
MW: 182,564 g/mol
Smältpunkt: 152 °C
Storage Temperature: Omgivande
MDL Number: MFCD18260051
CAS nummer: 1256833-56-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR