5-Chlorothieno[3,2-b]pyridine-6-carbaldehyde 99%
Leverantör: Apollo Scientific
Varning
OR55685-1G OR55685-250MG
APOSOR55685-1GEA 5075 SEK
APOSOR55685-1G APOSOR55685-250MG
5-Chlorothieno[3,2-b]pyridine-6-carbaldehyde 99%
5-Chlorothieno[3,2-b]pyridine-6-carbaldehyde
Formel: C₈H₄ClNOS
MW: 197,641 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD18838815
CAS nummer: 65977-56-2