2-Bromo-1-(4-cyclopropylphenyl)ethanone
Leverantör: Apollo Scientific
Fara
OR55672-1G OR55672-250MG
APOSOR55672-1GEA 1300 SEK
APOSOR55672-1G APOSOR55672-250MG
2-Bromo-1-(4-cyclopropylphenyl)ethanone
2-Bromo-1-(4-cyclopropylphenyl)ethanone
Formel: C₁₁H₁₁BrO
Storage Temperature: Frys
MDL Number: MFCD29040962
CAS nummer: 1934754-00-3
UN: 3265
ADR: 8,III