1-(4-Cyclopropylphenyl)ethanone
Leverantör: Apollo Scientific
Varning
OR55670-1G OR55670-250MG
APOSOR55670-1GEA 2200 SEK
APOSOR55670-1G APOSOR55670-250MG
1-(4-Cyclopropylphenyl)ethanone
1-(4-Cyclopropylphenyl)ethanone
Formel: C₁₁H₁₂O
MW: 160,212 g/mol
Storage Temperature: Frys
MDL Number: MFCD00102381
CAS nummer: 6921-45-5