4-Propylphenacyl bromide
Leverantör: Apollo Scientific
Fara
OR55608-1G OR55608-250MG OR55608-5G
APOSOR55608-1GEA 4100 SEK
APOSOR55608-1G APOSOR55608-250MG APOSOR55608-5G
4-Propylphenacyl bromide
4-Propylphenacyl bromide
Formel: C₁₁H₁₃BrO
MW: 241.124 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD11131363
CAS nummer: 64328-67-2
UN: 3261
ADR: 8 III