3-Amino-5-(methoxycarbonyl)benzeneboronic acid pinacol ester hydrochloride 95%
Leverantör: Apollo Scientific
Varning
Synonyms: Methyl 3-amino-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate hydrochloride
OR5553-1G OR5553-250MG
APOSOR5553-1GEA 4275 SEK
APOSOR5553-1G APOSOR5553-250MG
3-Amino-5-(methoxycarbonyl)benzeneboronic acid pinacol ester hydrochloride 95%
3-Amino-5-(methoxycarbonyl)benzeneboronic acid pinacol ester hydrochloride
Formel: C₁₄H₂₀BNO₄·ClH
MW: 313.585 g/mol
Smältpunkt: 125…127 °C
Storage Temperature: Omgivande
MDL Number: MFCD06659919
CAS nummer: 850567-50-9