2-(Aminomethyl)-3-bromopyridine hydrochloride
Leverantör: Apollo Scientific
Varning
Synonyms: (3-Bromopyridin-2-yl)methanamine hydrochloride
OR55277-1G OR55277-250MG OR55277-5G
APOSOR55277-1GEA 2650 SEK
APOSOR55277-1G APOSOR55277-250MG APOSOR55277-5G
2-(Aminomethyl)-3-bromopyridine hydrochloride
2-(Aminomethyl)-3-bromopyridine hydrochloride
Formel: C₆H₇BrN₂·ClH
MW: 223,498 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD20923742
CAS nummer: 1052271-58-5