1,3-Dimethyl-4-nitro-1H-pyrazole
Leverantör: Apollo Scientific
Varning
OR55208-1G OR55208-5G OR55208-25G
APOSOR55208-1GEA 360 SEK
APOSOR55208-1G APOSOR55208-5G APOSOR55208-25G
1,3-Dimethyl-4-nitro-1H-pyrazole
1,3-Dimethyl-4-nitro-1H-pyrazole
Formel: C₅H₇N₃O₂
MW: 141,128 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00464263
CAS nummer: 3920-38-5