4-Bromo-2-nitrothiophenol, Technical Grade
Leverantör: Apollo Scientific
Varning
Synonyms: 4-Bromo-2-nitrobenzenethiol
OR55173-1G OR55173-250MG OR55173-100MG
APOSOR55173-1GEA 3375 SEK
APOSOR55173-1G APOSOR55173-250MG APOSOR55173-100MG
4-Bromo-2-nitrothiophenol, Technical Grade
4-Bromo-2-nitrothiophenol
Formel: C₆H₄BrNO₂S
MW: 234,07 g/mol
Storage Temperature: Kylskåp
MDL Number: MFCD14652161
CAS nummer: 76209-02-4