5-Bromoimidazo[1,5-a]pyridine 95+%
Leverantör: Apollo Scientific
OR54716-1G OR54716-250MG
APOSOR54716-1GEA 3675 SEK
APOSOR54716-1G APOSOR54716-250MG
5-Bromoimidazo[1,5-a]pyridine 95+%
5-Bromoimidazo[1,5-a]pyridine
Formel: C₇H₅BrN₂
Storage Temperature: Kylskåp
MDL Number: MFCD06739219
CAS nummer: 885275-77-4