Piperidine-1-sulphonamide

Leverantör: Apollo Scientific

OR54585-1G OR54585-250MG
APOSOR54585-1GEA 11575 SEK
APOSOR54585-1G APOSOR54585-250MG
Piperidine-1-sulphonamide
Piperidine-1-sulphonamide
Formel: C₅H₁₂N₂O₂S
Storage Temperature: Omgivande
MDL Number: MFCD11171675
CAS nummer: 4108-90-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR