1-(Hydroxymethyl)cyclobutan-1-ol

Leverantör: Apollo Scientific
Fara

OR54527-1G OR54527-250MG
APOSOR54527-1GEA 14475 SEK
APOSOR54527-1G APOSOR54527-250MG
1-(Hydroxymethyl)cyclobutan-1-ol
1-(Hydroxymethyl)cyclobutan-1-ol
Formel: C₅H₁₀O₂
MW: 102,132 g/mol
Kokpunkt: 206…207 °C (756 mm Hg)
Densitet: 1,06 g/cm³ (25 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00045403
CAS nummer: 6970-72-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR