(3-Chlorocyclobutyl)methanol

Leverantör: Apollo Scientific
Varning

OR54434-1G OR54434-250MG
APOSOR54434-1GEA 3700 SEK
APOSOR54434-1G APOSOR54434-250MG
(3-Chlorocyclobutyl)methanol
(3-Chlorocyclobutyl)methanol
Formel: C₅H₉ClO
MW: 120,577 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD16659929
CAS nummer: 15963-47-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR