2-[4-(tert-Butoxycarbonyl)piperazin-1-yl]propanoic acid
Leverantör: Apollo Scientific
Varning
Synonyms: tert-Butyl 4-(1-carboxyethyl)piperazine-1-carboxylate, 4-(1-Carboxyethyl)piperazine, N1-BOC protected
OR5407-1G OR5407-5G
APOSOR5407-1GEA 1600 SEK
APOSOR5407-1G APOSOR5407-5G
2-[4-(tert-Butoxycarbonyl)piperazin-1-yl]propanoic acid
2-[4-(tert-Butoxycarbonyl)piperazin-1-yl]propanoic acid
Formel: C₁₂H₂₂N₂O₄
MW: 258,314 g/mol
Smältpunkt: 177 °C (dec.)
Storage Temperature: Omgivande
MDL Number: MFCD03410260
CAS nummer: 680579-19-5