5-Bromo-2-hydroxyphenylacetic acid

Leverantör: Apollo Scientific
Varning

Synonyms: 2-(5-Bromo-2-hydroxyphenyl)acetic acid

OR53099-1G OR53099-5G
APOSOR53099-1GEA 910 SEK
APOSOR53099-1G APOSOR53099-5G
5-Bromo-2-hydroxyphenylacetic acid
5-Bromo-2-hydroxyphenylacetic acid
Formel: C₈H₇BrO₃
MW: 231,043 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD16620636
CAS nummer: 38692-72-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR