3-Ethynyl-5-methylpyridine
Leverantör: Apollo Scientific
Varning
Synonyms: 5-Ethynyl-3-picoline, (5-Methylpyridin-3-yl)acetylene
OR53005-1G OR53005-250MG OR53005-100MG
APOSOR53005-1GEA 2975 SEK
APOSOR53005-1G APOSOR53005-250MG APOSOR53005-100MG
3-Ethynyl-5-methylpyridine
3-Ethynyl-5-methylpyridine
Formel: C₈H₇N
MW: 117.148 g/mol
Storage Temperature: Kylskåp
MDL Number: MFCD13175246
CAS nummer: 30413-53-7