Dimethyl (1,3-dihydro-3-oxo-1-isobenzofuranyl)phosphonate 98%
Leverantör: Apollo Scientific
Synonyms: Dimethyl (3-oxo-1,3-dihydroisobenzofuran-1-yl)phosphonate
OR52941-100G OR52941-25G OR52941-5G
APOSOR52941-100GEA 6475 SEK
APOSOR52941-100G APOSOR52941-25G APOSOR52941-5G
Dimethyl (1,3-dihydro-3-oxo-1-isobenzofuranyl)phosphonate 98%
Dimethyl (1,3-dihydro-3-oxo-1-isobenzofuranyl)phosphonate
Formel: C₁₀H₁₁O₅P
MW: 242,165 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD16883476
CAS nummer: 61260-15-9