cis-3-Aminocyclohexanecarboxylic acid hydrochloride 95%
Leverantör: Apollo Scientific
Varning
OR52453-5G OR52453-1G
APOSOR52453-5GEA 11975 SEK
APOSOR52453-5G APOSOR52453-1G
cis-3-Aminocyclohexanecarboxylic acid hydrochloride 95%
cis-3-Aminocyclohexanecarboxylic acid hydrochloride
Formel: C₇H₁₃NO₂·ClH
MW: 179,645 g/mol
Storage Temperature: Kylskåp
MDL Number: MFCD00787794
CAS nummer: 118785-96-9