2-Benzylbenzoic acid 98%

Leverantör: Apollo Scientific
Varning

Synonyms: α-Phenyl-o-toluic acid, 2-Carboxydiphenylmethane

OR5190-10G OR5190-25G
APOSOR5190-10GEA 1050 SEK
APOSOR5190-10G APOSOR5190-25G
2-Benzylbenzoic acid 98%
2-Benzylbenzoic acid
Formel: C₁₄H₁₂O₂
MW: 212,244 g/mol
Smältpunkt: 112…115 °C
Storage Temperature: Omgivande
MDL Number: MFCD00002484
CAS nummer: 612-35-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR