Naphthalene-1,3,5-trisulphonic acid 95%

Leverantör: Apollo Scientific
Fara

OR51841-25G OR51841-5G
APOSOR51841-25GEA 3025 SEK
APOSOR51841-25G APOSOR51841-5G
Naphthalene-1,3,5-trisulphonic acid 95%
Naphthalene-1,3,5-trisulphonic acid
Formel: C₁₀H₈O₉S₃
MW: 368,36 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD03265508
CAS nummer: 6654-64-4
UN: 2585
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR