2-(Aminomethyl)-5-hydroxypyridine hydrochloride 95%
Leverantör: Apollo Scientific
Varning
Synonyms: 6-(Aminomethyl)pyridin-3-ol hydrochloride, (5-Hydroxypyridin-2-yl)methylamine hydrochloride
OR51778-1G OR51778-250MG
APOSOR51778-1GEA 3175 SEK
APOSOR51778-1G APOSOR51778-250MG
2-(Aminomethyl)-5-hydroxypyridine hydrochloride 95%
2-(Aminomethyl)-5-hydroxypyridine hydrochloride
Formel: C₆H₈N₂O·ClH
MW: 160,601 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD17214179
CAS nummer: 1276056-66-6