1-Chloro-4-(2-phenylphenyl)benzene 99%
Leverantör: Apollo Scientific
OR51027-1G
OR51027-5G
APOSOR51027-1GEA
0
SEK
APOSOR51027-1G
APOSOR51027-5G
1-Chloro-4-(2-phenylphenyl)benzene 99%
1-Chloro-4-(2-phenylphenyl)benzene
Formel:
C₁₈H₁₃Cl MW: 264,749 g/mol Storage Temperature: Omgivande |
CAS nummer:
21711-54-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...