9-Phenyl-3,3′-bicarbazole 99%

Leverantör: Apollo Scientific

Synonyms: 9-Phenyl-3,3'-bicarbazolyl

OR51002-1G OR51002-5G
APOSOR51002-1GEA 855 SEK
APOSOR51002-1G APOSOR51002-5G
9-Phenyl-3,3′-bicarbazole 99%
9-Phenyl-3,3′-bicarbazole
Formel: C₃₀H₂₀N₂
MW: 408,493 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD22581311
CAS nummer: 1060735-14-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR