2,5,6,7-Tetrahydro-4H-indazol-4-one ≥97%

Leverantör: Apollo Scientific

OR510001-1G OR510001-250MG
APOSOR510001-1GEA 3300 SEK
APOSOR510001-1G APOSOR510001-250MG
2,5,6,7-Tetrahydro-4H-indazol-4-one ≥97%
2,5,6,7-Tetrahydro-4H-indazol-4-one
Formel: C₇H₈N₂O
MW: 136,151 g/mol
Storage Temperature: Kylskåp
CAS nummer: 912259-10-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR