2-(Methylamino)phenol

Leverantör: Apollo Scientific
Varning

Synonyms: 2-Hydroxy-N-methylaniline

OR5095-1G OR5095-5G
APOSOR5095-1GEA 875 SEK
APOSOR5095-1G APOSOR5095-5G
2-(Methylamino)phenol
2-(Methylamino)phenol
Formel: C₇H₉NO
MW: 123,152 g/mol
Smältpunkt: 89…94 °C
Storage Temperature: Omgivande
MDL Number: MFCD00462180
CAS nummer: 611-24-5
EINECS: 210-263-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR