2-Chloroquinoline-6-carbaldehyde 97%

Leverantör: Apollo Scientific
Varning

OR500027-100MG OR500027-250MG
APOSOR500027-100MGEA 1025 SEK
APOSOR500027-100MG APOSOR500027-250MG
2-Chloroquinoline-6-carbaldehyde 97%
2-Chloroquinoline-6-carbaldehyde
Formel: C₁₀H₆ClNO
Storage Temperature: Kylskåp
MDL Number: MFCD08236769
CAS nummer: 791626-59-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR