4-Aminocyclohexanecarbonitrile hydrochloride 97%
Leverantör: Apollo Scientific
Varning
OR500002-100MG OR500002-250MG
APOSOR500002-100MGEA 515 SEK
APOSOR500002-100MG APOSOR500002-250MG
4-Aminocyclohexanecarbonitrile hydrochloride 97%
4-Aminocyclohexanecarbonitrile hydrochloride
Formel: C₇H₁₂N₂·ClH
MW: 160,645 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD18914312
CAS nummer: 1303968-08-2