(2R)-2-(4-Hydroxyphenyl)propanoic acid 98%
Leverantör: Apollo Scientific
Varning
Synonyms: (R)-(4-Hydroxyphenyl)(methyl)acetic acid, (2R)-2-(4-Hydroxyphenyl)propionic acid, 4-[(1R)-1-Carboxyethyl]phenol
OR48264-1G OR48264-250MG OR48264-5G
APOSOR48264-1GEA 4800 SEK
APOSOR48264-1G APOSOR48264-250MG APOSOR48264-5G
(2R)-2-(4-Hydroxyphenyl)propanoic acid 98%
(2R)-2-(4-Hydroxyphenyl)propanoic acid
Formel: C₉H₁₀O₃
MW: 166,174 g/mol
Smältpunkt: 156…158 °C
Storage Temperature: Omgivande
MDL Number: MFCD21606316
CAS nummer: 59092-88-5