Methyl 5-formylpyrazine-2-carboxylate 96%
Leverantör: Apollo Scientific
OR480845-1G OR480845-250MG OR480845-5G
APOSOR480845-1GEA 2700 SEK
APOSOR480845-1G APOSOR480845-250MG APOSOR480845-5G
Methyl 5-formylpyrazine-2-carboxylate 96%
Methyl 5-formylpyrazine-2-carboxylate
Formel: C₇H₆N₂O₃
Storage Temperature: Omgivande
MDL Number: MFCD12755910
CAS nummer: 710322-57-9